Assay Details
Target Gene Details
Entrez Gene ID: | 327657 |
Gene Name: | serpin family A member 9 |
Gene Aliases: |
GCET1, SERPINA11, SERPINA11b |
Location: |
Chr.14:94462721-94476333 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINA9 | NM_001042518.1 | NP_001035983.1 | ||
| NM_001284275.1 | 2 | 577 | NP_001271204.1 | |
| NM_001284276.1 | 3 | 474 | NP_001271205.1 | |
| NM_175739.3 | 2 | 767 | NP_783866.2 | |
| XM_011536714.2 | 2 | 873 | XP_011535016.1 | |
| XM_011536715.2 | 3 | 737 | XP_011535017.1 | |
| XM_011536716.2 | 2 | 683 | XP_011535018.1 | |
| XM_011536717.1 | 2 | 1566 | XP_011535019.1 | |
| AK301082.1 | 2 | 661 | ||
| AY185496.1 | 2 | 767 | AAO32345.1 | |
| AY185497.1 | 2 | 577 | AAO32346.1 | |
| AY220118.1 | 2 | 681 | AAO65242.1 | |
| AY220119.1 | AAO65243.1 | |||
| AY220120.1 | 2 | 442 | AAO65244.1 | |
| AY220121.1 | 3 | 383 | AAO65245.1 | |
| AY358700.1 | 2 | 611 | AAQ89063.1 | |
| BC111498.1 | 2 | 670 | AAI11499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832863 | Chr.14:94348482 - 94501487 on Build GRCh38 | Gain |
|
| nsv510649 | Chr.14:94421103 - 94544514 on Build GRCh38 | Deletion |
|
| nsv470662 | Chr.14:94296542 - 94798793 on Build GRCh38 | Gain |
|
| nsv1407 | Chr.14:94450081 - 94477913 on Build GRCh38 | Insertion |
|
| nsv565623 | Chr.14:94393116 - 94635006 on Build GRCh38 | Gain |
|
| nsv832864 | Chr.14:94362101 - 94538150 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78283108] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map