Assay Details
Target Gene Details
Entrez Gene ID: | 9793 |
Gene Name: | cytoskeleton associated protein 5 |
Gene Aliases: |
CHTOG, MSPS, TOG, TOGp, ch-TOG |
Location: |
Chr.11:46743534-46846308 on Build GRCh38 |
Assay Gene Location: | Within Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CKAP5 | NM_001008938.3 | 39 | 5313 | NP_001008938.1 |
| NM_014756.3 | 38 | 5133 | NP_055571.2 | |
| BC035554.1 | 14 | 2096 | AAH35554.1 | |
| BC120869.1 | 38 | 5207 | ||
| BC120870.1 | 38 | 5205 | ||
| D43948.1 | 38 | 5234 | ||
| X92474.1 | 37 | 5033 | CAA63212.1 |
More Information
Set Membership: |
|

Genomic Map