Assay Details
Target Gene Details
Entrez Gene ID: | 54855 |
Gene Name: | family with sequence similarity 46 member C |
Gene Aliases: |
- |
Location: |
Chr.1:117605982-117628389 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM46C | NM_017709.3 | 2 | 3588 | NP_060179.2 |
| AK091327.1 | 1 | 771 | ||
| AL046016.1 | ||||
| BC036516.2 | ||||
| BC131726.1 | 2 | 3501 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007736 | Chr.1:117614642 - 117631600 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map