Assay Details
Target Gene Details
Entrez Gene ID: | 78989 |
Gene Name: | collectin subfamily member 11 |
Gene Aliases: |
3MC2, CL-K1-I, CL-K1-II, CL-K1-IIa, CL-K1-IIb, CLK1 |
Location: |
Chr.2:3594832-3644644 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COLEC11 | NM_001255982.1 | 2 | 415 | NP_001242911.1 |
| NM_001255983.1 | 2 | 415 | NP_001242912.1 | |
| NM_001255984.1 | 2 | 415 | NP_001242913.1 | |
| NM_001255985.1 | 3 | 305 | NP_001242914.1 | |
| NM_024027.4 | 2 | 415 | NP_076932.1 | |
| NM_199235.2 | 2 | 415 | NP_954705.1 | |
| NR_045659.1 | 2 | 415 | ||
| XM_005263853.4 | 2 | 109 | XP_005263910.1 | |
| XM_006711897.3 | 2 | 109 | XP_006711960.1 | |
| AB119525.1 | 2 | 301 | ||
| AB119650.1 | 2 | 301 | ||
| AB119651.1 | 2 | 301 | ||
| AB119652.1 | 2 | 301 | ||
| AK303824.1 | 3 | 305 | ||
| AK313840.1 | 2 | 417 | ||
| AY358439.1 | 2 | 118 | AAQ88805.1 | |
| BC000078.2 | 2 | 125 | AAH00078.1 | |
| BC009951.2 | 2 | 198 | AAH09951.1 | |
| BE259985.1 | 2 | 391 | ||
| BF316908.1 | 2 | 200 | ||
| BM547424.1 | 1 | 93 | ||
| CR936641.1 | CAI56781.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3702n100 | Chr.2:3361936 - 3733306 on Build GRCh38 | Gain |
|
| nsv468950 | Chr.2:3597839 - 3629432 on Build GRCh38 | Loss |
|
| nsv580822 | Chr.2:3552110 - 4305551 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map