Assay Details
Target Gene Details
Entrez Gene ID: | 949 |
Gene Name: | scavenger receptor class B member 1 |
Gene Aliases: |
CD36L1, CLA-1, CLA1, HDLQTL6, SR-BI, SRB1 |
Location: |
Chr.12:124777628-124863973 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCARB1 | NM_001082959.1 | 12 | 1994 | NP_001076428.1 |
| NM_005505.4 | 13 | 2123 | NP_005496.4 | |
| BC080647.1 | 13 | 1997 | AAH80647.1 | |
| CA428516.1 | 1 | 637 | ||
| Z22555.1 | 13 | 1939 | CAA80277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560595 | Chr.12:124737381 - 125019207 on Build GRCh38 | Gain |
|
| nsv560543 | Chr.12:124496040 - 124781090 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map