Assay Details
Target Gene Details
Entrez Gene ID: | 79802 |
Gene Name: | HHIP like 2 |
Gene Aliases: |
KIAA1822L |
Location: |
Chr.1:222501521-222548102 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HHIPL2 | NM_024746.3 | NP_079022.2 | ||
| XM_011509985.2 | XP_011508287.1 | |||
| XM_011509986.1 | XP_011508288.1 | |||
| XM_017002350.1 | XP_016857839.1 | |||
| AK023902.1 | BAB14717.1 | |||
| AY358602.1 | AAQ88965.1 | |||
| BC007638.1 | AAH07638.1 | |||
| BC015167.1 | AAH15167.1 | |||
| BC015201.1 | AAH15201.1 | |||
| BC016552.1 | AAH16552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160091 | Chr.1:221877143 - 222556663 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map