Assay Details
Target Gene Details
Entrez Gene ID: | 408 |
Gene Name: | arrestin beta 1 |
Gene Aliases: |
ARB1, ARR1 |
Location: |
Chr.11:75260122-75351831 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRB1 | NM_004041.4 | 16 | 2113 | NP_004032.2 |
| NM_020251.3 | 15 | 2089 | NP_064647.1 | |
| XM_011545034.2 | 17 | 1997 | XP_011543336.1 | |
| XM_011545035.2 | 16 | 1973 | XP_011543337.1 | |
| XM_017017750.1 | 18 | 2069 | XP_016873239.1 | |
| XM_017017751.1 | 17 | 2045 | XP_016873240.1 | |
| XM_017017752.1 | 17 | 2025 | XP_016873241.1 | |
| XM_017017753.1 | 17 | 2089 | XP_016873242.1 | |
| XM_017017754.1 | 16 | 2001 | XP_016873243.1 | |
| BC003636.2 | AAH03636.1 | |||
| CB853136.1 | 1 | 86 | ||
| CD365891.1 | 1 | 553 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832211 | Chr.11:75234794 - 75395081 on Build GRCh38 | Loss |
|
| dgv1225n100 | Chr.11:75226932 - 75348452 on Build GRCh38 | Gain |
|
| esv3626851 | Chr.11:75224463 - 75334026 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map