Assay Details
Target Gene Details
Entrez Gene ID: | 4154 |
Gene Name: | muscleblind like splicing regulator 1 |
Gene Aliases: |
EXP, MBNL |
Location: |
Chr.3:152243656-152465780 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBNL1 | XM_005247475.3 | XP_005247532.1 | ||
| XM_006713639.1 | XP_006713702.1 | |||
| XM_017006428.1 | XP_016861917.1 | |||
| XM_017006439.1 | XP_016861928.1 | |||
| AI971426.1 | ||||
| AW204030.1 |
Target Gene Details
Entrez Gene ID: | 401093 |
Gene Name: | MBNL1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:152262616-152269626 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBNL1-AS1 | NR_027037.1 | 2 | 1203 | |
| NR_027038.1 | 2 | 1214 | ||
| AK126417.1 | 1 | 300 | ||
| AK316515.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954493 | Chr.3:152196212 - 152409011 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map