Assay Details
Target Gene Details
Entrez Gene ID: | 162427 |
Gene Name: | family with sequence similarity 134 member C |
Gene Aliases: |
- |
Location: |
Chr.17:42579508-42609427 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM134C | NM_178126.3 | 9 | 2916 | NP_835227.1 |
| NR_026697.1 | 8 | 2885 | ||
| XM_011524440.2 | 9 | 2669 | XP_011522742.1 | |
| AK057409.1 | 1 | 755 | ||
| AK091125.1 | 8 | 3261 | ||
| AK225797.1 | 9 | 2864 | ||
| BC049370.1 | 9 | 2873 | AAH49370.1 | |
| BX647175.1 | 9 | 2807 | ||
| BX648069.1 | 1 | 1858 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map