Assay Details
Target Gene Details
Entrez Gene ID: | 6664 |
Gene Name: | SRY-box 11 |
Gene Aliases: |
MRD27 |
Location: |
Chr.2:5692667-5701385 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX11 | NM_003108.3 | 1 | 1185 | NP_003099.1 |
| AB028641.1 | 1 | 1185 | BAA88122.1 | |
| U23752.1 | 1 | 1205 | AAB08518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv676e199 | Chr.2:5689713 - 5694860 on Build GRCh38 | Deletion |
|
| nsv833292 | Chr.2:5663369 - 5827989 on Build GRCh38 | Gain+Loss |
|
| dgv609n67 | Chr.2:5665874 - 5710795 on Build GRCh38 | Gain |
|
| dgv608n67 | Chr.2:5647093 - 5731885 on Build GRCh38 | Gain |
|
| nsv828241 | Chr.2:5689145 - 5695527 on Build GRCh38 | Gain |
|
| nsv953442 | Chr.2:5690269 - 5697668 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map