Assay Details
Target Gene Details
Entrez Gene ID: | 55790 |
Gene Name: | chondroitin sulfate N-acetylgalactosaminyltransferase 1 |
Gene Aliases: |
CSGalNAcT-1, ChGn, beta4GalNAcT |
Location: |
Chr.8:19404161-19757940 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616532 | Chr.8:18974801 - 19484110 on Build GRCh38 | Gain |
|
| nsv610752 | Chr.8:19397305 - 19453985 on Build GRCh38 | Loss |
|
| nsv610751 | Chr.8:19368824 - 19427378 on Build GRCh38 | Loss |
|
| esv34007 | Chr.8:19395091 - 19624587 on Build GRCh38 | Loss |
|
| nsv522185 | Chr.8:19097292 - 20497802 on Build GRCh38 | Gain |
|
| nsv1016482 | Chr.8:18977297 - 19481976 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116759812] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map