Assay Details
Target Gene Details
Entrez Gene ID: | 731 |
Gene Name: | complement component 8 alpha subunit |
Gene Aliases: |
- |
Location: |
Chr.1:56854770-56918221 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8A | NM_000562.2 | 4 | 504 | NP_000553.1 |
| XM_011542079.2 | 4 | 478 | XP_011540381.1 | |
| XM_017002234.1 | 4 | 478 | XP_016857723.1 | |
| AK309596.1 | 4 | 478 | ||
| BC020702.1 | 4 | 477 | ||
| BC132911.1 | 4 | 396 | ||
| BC132913.1 | 4 | 396 | ||
| M16974.1 | 4 | 509 | AAA52200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508215 | Chr.1:56853787 - 56885477 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map