Assay Details
Target Gene Details
Entrez Gene ID: | 30848 |
Gene Name: | cancer/testis antigen 2 |
Gene Aliases: |
CAMEL, CT2, CT6.2, CT6.2a, CT6.2b, ESO2, LAGE-1, LAGE2B |
Location: |
Chr.X:154651972-154653579 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTAG2 | NM_020994.4 | 2 | 488 | NP_066274.2 |
| NM_172377.4 | NP_758965.2 | |||
| AJ012833.1 | CAA10193.1 | |||
| AJ012834.1 | CAA10194.1 | |||
| AJ012835.1 | 2 | 479 | CAA10196.1 | |
| AJ223040.1 | 2 | 488 | CAA11043.1 | |
| AJ223041.1 | CAA11044.1 | |||
| AY679088.1 | ||||
| AY679089.1 | ||||
| BC002833.2 | 2 | 442 | AAH02833.1 | |
| BC113998.1 | ||||
| BC114933.1 | ||||
| BC128045.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518228 | Chr.X:154601444 - 154719706 on Build GRCh38 | Gain |
|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map