Assay Details
Target Gene Details
Entrez Gene ID: | 57134 |
Gene Name: | mannosidase alpha class 1C member 1 |
Gene Aliases: |
HMIC, MAN1A3, MAN1C, pp6318 |
Location: |
Chr.1:25616820-25786207 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAN1C1 | NM_001289010.1 | NP_001275939.1 | ||
| NM_020379.3 | NP_065112.1 | |||
| XM_017001860.1 | XP_016857349.1 | |||
| XM_017001861.1 | XP_016857350.1 | |||
| AB209275.1 | BAD92512.1 | |||
| AF261655.1 | AAF97058.1 | |||
| BC137017.1 |
More Information
Set Membership: |
|

Genomic Map