Assay Details
Target Gene Details
Entrez Gene ID: | 163479 |
Gene Name: | fibronectin type III domain containing 7 |
Gene Aliases: |
- |
Location: |
Chr.1:108712934-108742749 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNDC7 | NM_001144937.1 | 8 | 1557 | NP_001138409.1 |
| AK093157.1 | 4 | 883 | BAC04077.1 | |
| BC057762.1 | 4 | 930 | AAH57762.1 | |
| BC130421.1 | 4 | 883 |

Genomic Map