Assay Details
Target Gene Details
Entrez Gene ID: | 2872 |
Gene Name: | MAP kinase interacting serine/threonine kinase 2 |
Gene Aliases: |
GPRK7, MNK2 |
Location: |
Chr.19:2037471-2051244 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MKNK2 | NM_017572.3 | 11 | 1090 | NP_060042.2 |
| NM_199054.2 | 11 | 1090 | NP_951009.1 | |
| AB209074.1 | 3 | 261 | BAD92311.1 | |
| AF125532.1 | 10 | 867 | AAF17226.1 | |
| AF237775.1 | 10 | 881 | AAG26336.1 | |
| AF237776.1 | 10 | 881 | AAG26337.1 | |
| AK001730.1 | 1 | 141 | BAA91866.1 | |
| AK092536.1 | ||||
| AK293742.1 | 9 | 749 | ||
| AK303393.1 | 8 | 760 | ||
| AL137565.1 | 7 | 552 | CAB70816.1 | |
| AL513974.3 | 1 | 201 | ||
| BC018345.1 | 3 | 259 | AAH18345.1 | |
| BC073140.1 | 11 | 1070 | AAH73140.1 | |
| BM977403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss |
|
| nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss |
|
| nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map