Assay Details
Target Gene Details
Entrez Gene ID: | 132864 |
Gene Name: | cytoplasmic polyadenylation element binding protein 2 |
Gene Aliases: |
CPE-BP2, CPEB-2, hCPEB-2 |
Location: |
Chr.4:15002451-15070153 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB2 | NM_001177381.1 | 11 | 3024 | NP_001170852.1 |
| NM_001177382.1 | 12 | 3105 | NP_001170853.1 | |
| NM_001177383.1 | 11 | 3015 | NP_001170854.1 | |
| NM_001177384.1 | 10 | 3000 | NP_001170855.1 | |
| NM_182485.2 | 11 | 3081 | NP_872291.2 | |
| NM_182646.2 | 10 | 2991 | NP_872587.2 | |
| XM_005248135.3 | 12 | 3337 | XP_005248192.2 | |
| XM_011513777.2 | 11 | 3313 | XP_011512079.1 | |
| XM_017007733.1 | 12 | 1473 | XP_016863222.1 | |
| XM_017007734.1 | 12 | 1474 | XP_016863223.1 | |
| AY247744.1 | 11 | 1857 | AAP37585.1 | |
| AY255519.1 | 10 | 1767 | AAP41553.1 | |
| BC103939.1 | 12 | 1696 | ||
| BC103940.1 | 11 | 1819 | AAI03941.1 | |
| BC103941.1 | 11 | 1756 | AAI03942.1 | |
| BC103942.1 | 13 | 1777 | AAI03943.1 | |
| BC105925.1 | 10 | 1738 | AAI05926.1 | |
| BU854092.1 | 1 | 54 | ||
| BX640753.1 | 2 | 258 | CAE45860.1 |
Target Gene Details
Entrez Gene ID: | 101929095 |
Gene Name: | uncharacterized LOC101929095 |
Gene Aliases: |
- |
Location: |
Chr.4:15004942-15427914 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929095 | NR_125911.1 | |||
| DB497821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829868 | Chr.4:14962935 - 15163082 on Build GRCh38 | Gain |
|
| nsv461280 | Chr.4:14960872 - 15079750 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map