Assay Details
Target Gene Details
Entrez Gene ID: | 6804 |
Gene Name: | syntaxin 1A |
Gene Aliases: |
HPC-1, P35-1, STX1, SYN1A |
Location: |
Chr.7:73699205-73719702 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX1A | NM_001165903.1 | 10 | 1999 | NP_001159375.1 |
| NM_004603.3 | 10 | 2045 | NP_004594.1 | |
| AI951874.1 | 1 | 94 | ||
| BC003011.1 | 10 | 1971 | AAH03011.1 | |
| BC064644.1 | 10 | 2016 | AAH64644.1 | |
| D37932.1 | 10 | 1989 | BAA07151.1 | |
| L37792.1 | 10 | 1990 | AAA53519.1 | |
| U12918.1 | 9 | 1948 | AAA20940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831028 | Chr.7:73556313 - 73816595 on Build GRCh38 | Loss |
|
| nsv469841 | Chr.7:73621181 - 73770495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map