Assay Details
Target Gene Details
Entrez Gene ID: | 353116 |
Gene Name: | Rab interacting lysosomal protein like 1 |
Gene Aliases: |
GOSPEL, RLP1 |
Location: |
Chr.12:123471073-123533718 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RILPL1 | NM_001319243.1 | 8 | 1798 | NP_001306172.1 |
| NM_001319244.1 | 7 | 1259 | NP_001306173.1 | |
| NM_001319302.1 | 8 | 2209 | NP_001306231.1 | |
| NM_178314.4 | 7 | 1710 | NP_847884.2 | |
| XM_006719372.2 | 8 | 1728 | XP_006719435.1 | |
| XM_006719373.3 | 7 | 1081 | XP_006719436.1 | |
| XM_011538282.2 | 8 | 1769 | XP_011536584.1 | |
| XM_011538284.2 | 7 | 1300 | XP_011536586.1 | |
| XM_011538285.2 | 7 | 1061 | XP_011536587.1 | |
| AK096697.1 | 9 | 1640 | BAC04845.1 | |
| BC080626.1 | 7 | 1259 | AAH80626.1 | |
| BC089444.1 | 7 | 1512 | AAH89444.1 |
Target Gene Details
Entrez Gene ID: | 11066 |
Gene Name: | small nuclear ribonucleoprotein U11/U12 subunit 35 |
Gene Aliases: |
HM-1, U1SNRNPBP |
Location: |
Chr.12:123457644-123472362 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRNP35 | NR_104103.1 | 2 | 1298 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037260 | Chr.12:123472202 - 123538522 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map