Assay Details
Target Gene Details
Entrez Gene ID: | 5929 |
Gene Name: | RB binding protein 5, histone lysine methyltransferase complex subunit |
Gene Aliases: |
RBQ3, SWD1 |
Location: |
Chr.1:205086142-205122022 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBBP5 | NM_001193272.1 | 14 | 3593 | NP_001180201.1 |
| NM_001193273.1 | 14 | 3648 | NP_001180202.1 | |
| NM_005057.3 | 14 | 3707 | NP_005048.2 | |
| XM_011509833.2 | 14 | 3702 | XP_011508135.1 | |
| BC037284.2 | 14 | 3546 | AAH37284.1 | |
| BG392257.1 | ||||
| CB133829.1 | 1 | 442 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819953 | Chr.1:205086869 - 205087412 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map