Assay Details
Target Gene Details
Entrez Gene ID: | 64838 |
Gene Name: | fibronectin type III domain containing 4 |
Gene Aliases: |
FRCP1 |
Location: |
Chr.2:27491883-27495303 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNDC4 | NM_022823.2 | NP_073734.1 | ||
| XM_005264499.3 | XP_005264556.1 | |||
| AK026015.1 | BAB15321.1 | |||
| AY358136.1 | AAQ88503.1 | |||
| BC032725.1 | AAH32725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv817684 | Chr.2:27434300 - 27623736 on Build GRCh38 | Gain |
|

Genomic Map