Assay Details
Target Gene Details
Entrez Gene ID: | 83931 |
Gene Name: | serine/threonine kinase 40 |
Gene Aliases: |
SHIK, SgK495 |
Location: |
Chr.1:36339619-36385927 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK40 | NM_001282546.1 | 11 | 3314 | NP_001269475.1 |
| NM_001282547.1 | 11 | 3299 | NP_001269476.1 | |
| NM_032017.2 | 12 | 3536 | NP_114406.1 | |
| AK000808.1 | 1 | 1221 | ||
| AK024504.1 | 9 | 4488 | BAB15794.1 | |
| AK075029.1 | 9 | 2287 | BAC11361.1 | |
| AK075048.1 | 12 | 2824 | BAC11371.1 | |
| AK131560.1 | 3 | 3816 | BAD18694.1 | |
| AL834137.1 | 12 | 3493 | CAD38852.1 | |
| AW269508.1 | 1 | 364 | ||
| AY198395.1 | 11 | 3269 | AAP20040.1 | |
| BC005169.2 | 6 | 2498 | AAH05169.3 | |
| BC007835.2 | 11 | 3267 | AAH07835.2 | |
| BC008344.1 | 11 | 3275 | AAH08344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950610 | Chr.1:36326200 - 36350399 on Build GRCh38 | Deletion |
|
| nsv524394 | Chr.1:36323945 - 36382165 on Build GRCh38 | Loss |
|
| nsv834702 | Chr.1:36190630 - 36391731 on Build GRCh38 | Loss |
|
| esv2758931 | Chr.1:36154538 - 36421487 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map