Assay Details
Target Gene Details
Entrez Gene ID: | 23157 |
Gene Name: | septin 6 |
Gene Aliases: |
SEP2, SEPT2 |
Location: |
Chr.X:119615724-119693370 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPT6 | NM_145800.3 | 11 | 2109 | NP_665799.1 |
| NM_145802.3 | 10 | 2064 | NP_665801.1 | |
| XM_005262400.3 | 11 | 5243 | XP_005262457.1 | |
| XM_006724748.2 | 10 | 5181 | XP_006724811.1 | |
| XM_006724750.2 | 11 | 3961 | XP_006724813.1 | |
| XM_011531317.2 | 11 | 1934 | XP_011529619.1 | |
| XM_011531318.2 | 11 | 3978 | XP_011529620.1 | |
| AF397023.1 | 10 | 1909 | AAN76547.1 | |
| AK026589.1 | 11 | 1902 | ||
| AY679521.1 | 10 | 1882 | AAT80339.1 | |
| BC011922.2 | 10 | 1852 | AAH11922.3 |
More Information
Set Membership: |
|

Genomic Map