Assay Details
Target Gene Details
Entrez Gene ID: | 10380 |
Gene Name: | 3'(2'), 5'-bisphosphate nucleotidase 1 |
Gene Aliases: |
HEL20, PIP |
Location: |
Chr.1:220057482-220089916 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPNT1 | NM_001286149.1 | 8 | 881 | NP_001273078.1 |
| NM_001286150.1 | 8 | 901 | NP_001273079.1 | |
| NM_001286151.1 | 8 | 923 | NP_001273080.1 | |
| NM_006085.5 | 9 | 1009 | NP_006076.4 | |
| XM_005272998.2 | 10 | 1025 | XP_005273055.1 | |
| XM_005272999.4 | 10 | 1134 | XP_005273056.1 | |
| XM_005273000.4 | 9 | 1058 | XP_005273057.1 | |
| XM_005273004.2 | 8 | 782 | XP_005273061.1 | |
| XM_005273005.4 | 8 | 894 | XP_005273062.1 | |
| XM_006711114.3 | 9 | 998 | XP_006711177.1 | |
| XM_011509063.2 | 9 | 972 | XP_011507365.1 | |
| XM_011509064.2 | 8 | 881 | XP_011507366.1 | |
| XM_017000043.1 | 8 | 973 | XP_016855532.1 | |
| XM_017000044.1 | 7 | 808 | XP_016855533.1 | |
| XM_017000046.1 | 7 | 732 | XP_016855535.1 | |
| AF125042.1 | 9 | 946 | AAD17329.1 | |
| AJ249339.1 | 9 | 974 | CAB65115.1 | |
| AK021941.1 | 4 | 407 | ||
| AK291943.1 | 9 | 944 | ||
| AK298476.1 | 8 | 826 | ||
| AK300777.1 | 8 | 846 | ||
| AK315962.1 | 8 | 903 | ||
| BC017801.1 | AAH17801.1 | |||
| EU794594.1 | 9 | 946 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832636 | Chr.1:219966355 - 220121010 on Build GRCh38 | Loss |
|
| nsv4632 | Chr.1:220041527 - 220076852 on Build GRCh38 | Insertion |
|
| nsv549222 | Chr.1:220053390 - 220073050 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map