Assay Details
Target Gene Details
Entrez Gene ID: | 5093 |
Gene Name: | poly(rC) binding protein 1 |
Gene Aliases: |
HEL-S-85, HNRPE1, HNRPX, hnRNP-E1, hnRNP-X |
Location: |
Chr.2:70087453-70089203 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCBP1 | NM_006196.3 | 1 | 524 | NP_006187.2 |
| AK130439.1 | 1 | 205 | ||
| AK314225.1 | 1 | 391 | ||
| BC039742.1 | 1 | 385 | AAH39742.1 | |
| BM794597.1 | ||||
| FJ224329.1 | 1 | 385 | ||
| U24223.1 | 1 | 410 | AAA91317.1 | |
| X78137.1 | 1 | 263 | CAA55016.1 | |
| Z29505.1 | 1 | 318 | CAA82631.1 |
More Information
Set Membership: |
|

Genomic Map