Assay Details
Target Gene Details
Entrez Gene ID: | 8394 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase type 1 alpha |
Gene Aliases: |
- |
Location: |
Chr.1:151198003-151249536 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map