Assay Details
Target Gene Details
Entrez Gene ID: | 83643 |
Gene Name: | coiled-coil domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.10:12896625-13099773 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC3 | NM_001282658.1 | 7 | 2749 | NP_001269587.1 |
| NM_031455.3 | 3 | 2184 | NP_113643.1 | |
| AF190154.1 | 1 | 748 | AAQ13698.1 | |
| AK024578.1 | 1 | 1167 | ||
| AK095792.1 | 7 | 2628 | ||
| AK123393.1 | 1 | 2325 | ||
| AK124195.1 | 1 | 1887 | ||
| AL136562.1 | 3 | 2100 | CAB66497.1 | |
| BC051334.1 | 3 | 2184 | AAH51334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5831 | Chr.10:12862255 - 12897166 on Build GRCh38 | Insertion |
|
| nsv971901 | Chr.10:12897050 - 12903518 on Build GRCh38 | Duplication |
|

Genomic Map