Assay Details
Target Gene Details
Entrez Gene ID: | 582 |
Gene Name: | Bardet-Biedl syndrome 1 |
Gene Aliases: |
BBS2L2 |
Location: |
Chr.11:66510648-66533613 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBS1 | NM_024649.4 | 4 | 262 | NP_078925.3 |
| AF503941.1 | 4 | 262 | AAM92770.1 | |
| AK095638.1 | 3 | 269 | ||
| AK294962.1 | ||||
| AK296311.1 | 4 | 268 | ||
| AK302657.1 | 3 | 240 | ||
| BC047642.1 | 4 | 268 | AAH47642.1 | |
| BC109064.2 | 4 | 255 | AAI09065.1 | |
| BC109065.1 | 4 | 255 | AAI09066.1 | |
| BX647612.1 | 4 | 291 | CAI46060.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map