Assay Details
Target Gene Details
Entrez Gene ID: | 10786 |
Gene Name: | solute carrier family 17 member 3 |
Gene Aliases: |
GOUT4, NPT4, UAQTL4 |
Location: |
Chr.6:25845100-25874243 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A3 | NM_001098486.1 | 10 | 1297 | NP_001091956.1 |
| NM_006632.3 | 9 | 1063 | NP_006623.2 | |
| AK296070.1 | ||||
| AK298271.1 | 10 | 1297 | ||
| AK298297.1 | ||||
| BC017952.1 | 9 | 1032 | AAH17952.1 | |
| CB163247.1 | 4 | 435 | ||
| U90545.1 | 9 | 1272 | AAB53423.1 |
More Information
Set Membership: |
|

Genomic Map