Assay Details
Target Gene Details
Entrez Gene ID: | 9459 |
Gene Name: | Rac/Cdc42 guanine nucleotide exchange factor 6 |
Gene Aliases: |
COOL2, Cool-2, MRX46, PIXA, alpha-PIX, alphaPIX |
Location: |
Chr.X:136665547-136781656 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF6 | NM_001306177.1 | 21 | 4652 | NP_001293106.1 |
| NM_004840.2 | 22 | 5114 | NP_004831.1 | |
| XM_005262499.2 | 20 | 5279 | XP_005262556.1 | |
| XM_011531412.2 | 23 | 5507 | XP_011529714.1 | |
| XM_011531413.1 | 22 | 5438 | XP_011529715.1 | |
| XM_011531414.1 | 21 | 5417 | XP_011529716.1 | |
| XM_011531415.2 | XP_011529717.1 | |||
| XM_011531416.2 | XP_011529718.1 | |||
| XM_011531417.1 | 21 | 5056 | XP_011529719.1 | |
| XM_017029975.1 | 23 | 5519 | XP_016885464.1 | |
| XM_017029976.1 | 19 | 4897 | XP_016885465.1 | |
| AK129909.1 | 1 | 1572 | ||
| BC033768.1 | 10 | 3285 | AAH33768.1 | |
| BC039856.1 | 22 | 5114 | AAH39856.1 | |
| BC043505.1 | 21 | 4618 | AAH43505.1 | |
| BX537390.1 | 22 | 4719 | CAD97632.1 | |
| D25304.1 | 22 | 4645 |
More Information
Set Membership: |
|

Genomic Map