Assay Details
Target Gene Details
Entrez Gene ID: | 2243 |
Gene Name: | fibrinogen alpha chain |
Gene Aliases: |
Fib2 |
Location: |
Chr.4:154583126-154590766 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGA | NM_000508.4 | NP_000499.1 | ||
| NM_021871.3 | NP_068657.1 | |||
| AK290559.1 | ||||
| BC020764.1 | AAH20764.1 | |||
| BC070246.1 | AAH70246.1 | |||
| BC098280.1 | AAH98280.1 | |||
| BC099706.1 | AAH99706.1 | |||
| BC099720.1 | AAH99720.1 | |||
| BC101935.1 | AAI01936.1 | |||
| BC105803.1 | AAI05804.1 | |||
| BG565064.1 | ||||
| BX494953.1 | ||||
| CB122305.1 | ||||
| CN393696.1 | ||||
| J00127.1 | AAA52426.1 | |||
| J00128.1 | AAA52427.1 | |||
| K02272.1 | AAA52428.1 | |||
| KU178062.1 | ||||
| KU178063.1 | ||||
| KU178064.1 | ||||
| KU178065.1 | ||||
| M58569.1 | AAC97142.1 |
More Information
Set Membership: |
|

Genomic Map