Assay Details
Target Gene Details
Entrez Gene ID: | 5554 |
Gene Name: | proline rich protein HaeIII subfamily 1 |
Gene Aliases: |
Db-s, PA, PIF-S, PRH2, Pr1/Pr2 |
Location: |
Chr.12:10880961-11171623 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRH1 | NM_001291314.1 | NP_001278243.1 | ||
| NM_001291315.1 | NP_001278244.1 | |||
| AB593029.1 | ||||
| BI772091.1 | ||||
| BM476484.1 | ||||
| BU543118.1 |
Target Gene Details
Entrez Gene ID: | 100533464 |
Gene Name: | PRH1-PRR4 readthrough |
Gene Aliases: |
PRH1 |
Location: |
Chr.12:10845849-11171625 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRH1-PRR4 | NR_037918.2 | |||
| AB593030.1 |
Target Gene Details
Entrez Gene ID: | 5555 |
Gene Name: | proline rich protein HaeIII subfamily 2 |
Gene Aliases: |
PRP-1/PRP-2, Pr, pr1/Pr2 |
Location: |
Chr.12:10929236-10934845 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRH2 | NM_001110213.1 | 4 | 1125 | NP_001103683.1 |
| BX538206.1 | 5 | 1075 | ||
| BX538207.1 | 5 | 1061 | ||
| BX641094.1 | 3 | 2274 | CAE46044.1 | |
| BX647085.1 | 5 | 957 | ||
| BX647155.1 | 4 | 2168 | ||
| BX647156.1 | 5 | 1029 | ||
| BX648436.1 | 5 | 1049 | ||
| BX648656.1 | 3 | 2200 | ||
| BX648871.1 | 2 | 2245 | ||
| BX648901.1 | 4 | 1055 | ||
| CR749583.1 | 5 | 888 | CAH18378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832330 | Chr.12:10925079 - 11054358 on Build GRCh38 | Gain |
|
| esv2759879 | Chr.12:10817021 - 11561987 on Build GRCh38 | Loss |
|
| nsv1038277 | Chr.12:10908379 - 11351157 on Build GRCh38 | Loss |
|
| nsv428272 | Chr.12:10917246 - 11561987 on Build GRCh38 | Gain+Loss |
|
| esv3628550 | Chr.12:10925203 - 10950111 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map