Assay Details
Target Gene Details
Entrez Gene ID: | 5875 |
Gene Name: | Rab geranylgeranyltransferase alpha subunit |
Gene Aliases: |
PTAR3 |
Location: |
Chr.14:24265538-24271627 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RABGGTA | NM_004581.5 | 1 | 283 | NP_004572.3 |
| NM_182836.2 | NP_878256.1 | |||
| AK291347.1 | ||||
| AK292613.1 | ||||
| AK308715.1 | ||||
| AL547259.3 | ||||
| AU125148.1 | 1 | 193 | ||
| BC003093.1 | AAH03093.1 | |||
| CB995791.1 | ||||
| DA379718.1 | ||||
| DB116988.1 | 1 | 234 | ||
| Y08200.1 | 1 | 46 | CAA69382.1 |
More Information
Set Membership: |
|

Genomic Map