Assay Details
Target Gene Details
Entrez Gene ID: | 10949 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A0 |
Gene Aliases: |
HNRPA0 |
Location: |
Chr.5:137751384-137754350 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPA0 | NM_006805.3 | 1 | 321 | NP_006796.1 |
| BC001008.1 | 1 | 320 | AAH01008.1 | |
| BC007271.1 | 1 | 49 | AAH07271.1 | |
| BC009284.2 | 1 | 93 | AAH09284.1 | |
| BC011972.1 | 1 | 312 | AAH11972.1 | |
| BC012980.2 | 1 | 320 | AAH12980.1 | |
| BC018949.2 | 1 | 320 | AAH18949.1 | |
| BC019271.1 | 1 | 300 | AAH19271.1 | |
| BC028976.1 | 1 | 321 | AAH28976.1 | |
| BC030249.1 | 1 | 321 | AAH30249.1 | |
| CR456986.1 | CAG33267.1 | |||
| U23803.1 | 1 | 301 | AAA65094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830500 | Chr.5:137672066 - 137845314 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map