Assay Details
Target Gene Details
Entrez Gene ID: | 80150 |
Gene Name: | asparaginase like 1 |
Gene Aliases: |
ALP, ALP1, CRASH |
Location: |
Chr.11:62337302-62401399 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASRGL1 | NM_001083926.1 | 2 | 448 | NP_001077395.1 |
| NM_025080.3 | 2 | 371 | NP_079356.3 | |
| XM_005274305.3 | 2 | 231 | XP_005274362.1 | |
| XM_005274306.2 | 2 | 293 | XP_005274363.1 | |
| XM_011545265.2 | 2 | 330 | XP_011543567.1 | |
| XM_011545266.2 | 2 | 330 | XP_011543568.1 | |
| XM_017018354.1 | 2 | 330 | XP_016873843.1 | |
| AF411076.1 | 2 | 180 | AAM28434.1 | |
| AK090908.1 | 2 | 220 | ||
| AK091894.1 | 2 | 302 | ||
| AK225802.1 | 2 | 139 | ||
| AK297158.1 | 2 | 222 | ||
| AK309845.1 | 2 | 216 | ||
| AK313069.1 | 2 | 231 | ||
| BC021295.2 | 2 | 169 | ||
| BC064963.1 | 2 | 204 | AAH64963.1 | |
| BC093070.1 | 2 | 273 | AAH93070.1 | |
| BP308677.1 | 2 | 371 | ||
| DA159051.1 | 2 | 302 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053587 | Chr.11:62326400 - 62390144 on Build GRCh38 | Gain |
|
| esv3626599 | Chr.11:62325417 - 62389620 on Build GRCh38 | Gain |
|
| esv3626600 | Chr.11:62326560 - 62354647 on Build GRCh38 | Gain |
|
| nsv516460 | Chr.11:62337465 - 62386689 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map