Assay Details
Target Gene Details
Entrez Gene ID: | 51474 |
Gene Name: | LIM domain and actin binding 1 |
Gene Aliases: |
EPLIN, SREBP3 |
Location: |
Chr.12:50175780-50283570 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIMA1 | NM_001113546.1 | 11 | 2310 | NP_001107018.1 |
| NM_001113547.1 | 8 | 2191 | NP_001107019.1 | |
| NM_001243775.1 | 5 | 1918 | NP_001230704.1 | |
| NM_016357.4 | 11 | 2307 | NP_057441.1 | |
| XM_011538455.2 | 11 | 2370 | XP_011536757.1 | |
| XM_017019429.1 | 6 | 1820 | XP_016874918.1 | |
| XM_017019430.1 | 11 | 2661 | XP_016874919.1 | |
| AB209512.1 | 11 | 2246 | BAD92749.1 | |
| AF157325.1 | 5 | 1917 | AAF67491.1 | |
| AF198454.1 | 11 | 2234 | AAF23755.1 | |
| AF198455.1 | 8 | 2129 | AAF23756.1 | |
| AF218025.1 | 7 | 1442 | AAG17267.1 | |
| AK000057.1 | 5 | 1233 | BAA90914.1 | |
| AK000335.1 | 8 | 1831 | BAA91092.1 | |
| AK000372.1 | 11 | 2217 | BAA91120.1 | |
| AK023649.1 | 5 | 1906 | BAB14625.1 | |
| AK096172.1 | 5 | 2498 | ||
| AK222971.1 | 11 | 2230 | BAD96691.1 | |
| AK297766.1 | 9 | 1903 | ||
| AK314447.1 | 11 | 2219 | ||
| AL136911.1 | 11 | 2226 | CAB66845.1 | |
| BC001247.1 | 8 | 1836 | AAH01247.2 | |
| BC010664.1 | 1 | 684 | AAH10664.1 | |
| BC110815.1 | 11 | 2244 | AAI10816.1 | |
| BC136763.1 | 11 | 2193 |
More Information
Set Membership: |
|

Genomic Map