Assay Details
Target Gene Details
Entrez Gene ID: | 3958 |
Gene Name: | lectin, galactoside binding soluble 3 |
Gene Aliases: |
CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2 |
Location: |
Chr.14:55129217-55145430 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGALS3 | NM_001177388.1 | NP_001170859.1 | ||
| NM_002306.3 | 6 | 781 | NP_002297.2 | |
| NR_003225.2 | 4 | 1737 | ||
| XM_011536759.2 | 7 | 771 | XP_011535061.2 | |
| AB006780.1 | 6 | 746 | BAA22164.1 | |
| AB209391.1 | 5 | 4824 | BAD92628.1 | |
| AF266280.1 | 4 | 1064 | AAG44701.1 | |
| AK314929.1 | 6 | 780 | ||
| BC001120.2 | 6 | 717 | AAH01120.1 | |
| BC053667.1 | 6 | 732 | AAH53667.1 | |
| BC068068.1 | 6 | 727 | AAH68068.1 | |
| BG332134.1 | ||||
| BU681070.1 | 1 | 237 | ||
| BX161455.1 | 4 | 1532 | CAD61918.1 | |
| BX641090.1 | 4 | 1738 | CAE46042.1 | |
| CD518618.1 | 6 | 781 | ||
| CR456897.1 | 5 | 693 | CAG33178.1 | |
| CR542097.1 | 5 | 693 | CAG46894.1 | |
| M35368.1 | 6 | 715 | AAA88086.1 | |
| M36682.1 | 6 | 726 | AAA36163.1 | |
| M57710.1 | 5 | 711 | AAA35607.1 | |
| M64303.1 | 5 | 711 | ||
| S59012.1 | 5 | 693 | AAB26229.1 |
More Information
Set Membership: |
|

Genomic Map