Assay Details
Target Gene Details
Entrez Gene ID: | 1948 |
Gene Name: | ephrin B2 |
Gene Aliases: |
EPLG5, HTKL, Htk-L, LERK5 |
Location: |
Chr.13:106489731-106535040 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFNB2 | NM_004093.3 | 5 | 2079 | NP_004084.1 |
| XM_017020406.1 | 5 | 10340 | XP_016875895.1 | |
| BC105955.1 | 5 | 2024 | AAI05956.1 | |
| BC105956.1 | 5 | 2024 | AAI05957.1 | |
| BC105957.1 | 5 | 2024 | AAI05958.1 | |
| BQ433342.1 | 1 | 95 | ||
| U81262.1 | 5 | 2010 | AAD03786.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633458 | Chr.13:106127915 - 106528334 on Build GRCh38 | Gain |
|
| nsv1039628 | Chr.13:106180176 - 106649540 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75769989] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map