Assay Details
Target Gene Details
Entrez Gene ID: | 29114 |
Gene Name: | transgelin 3 |
Gene Aliases: |
NP22, NP24, NP25 |
Location: |
Chr.3:111998739-112013888 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAGLN3 | NM_001008272.1 | 1 | 113 | NP_001008273.1 |
| NM_013259.2 | 1 | 113 | NP_037391.2 | |
| AF087855.1 | AAP97165.1 | |||
| AF303058.1 | AAL09330.1 | |||
| AK315289.1 | ||||
| BI752099.1 | 1 | 188 | ||
| BI919263.1 | 1 | 181 |
More Information
Set Membership: |
|

Genomic Map