Assay Details
Target Gene Details
Entrez Gene ID: | 117157 |
Gene Name: | SH2 domain containing 1B |
Gene Aliases: |
EAT2 |
Location: |
Chr.1:162395266-162412138 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH2D1B | NM_053282.4 | NP_444512.2 | ||
| AF484964.1 | AAM28522.1 | |||
| AL709120.1 | ||||
| BC022407.1 | AAH22407.1 | |||
| BC066595.1 | AAH66595.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758973 | Chr.1:162094678 - 162533566 on Build GRCh38 | Gain |
|
| nsv516482 | Chr.1:162138329 - 162905405 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map