Assay Details
Target Gene Details
Entrez Gene ID: | 7571 |
Gene Name: | zinc finger protein 23 |
Gene Aliases: |
KOX16, ZNF359, ZNF612, Zfp612 |
Location: |
Chr.16:71447597-71462252 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF23 | NM_001304492.1 | NP_001291421.1 | ||
| NM_001304493.1 | NP_001291422.1 | |||
| NM_001304494.1 | NP_001291423.1 | |||
| NM_145911.2 | 1 | 193 | NP_666016.1 | |
| AF507946.1 | 1 | 184 | AAM33359.1 | |
| AK091039.1 | ||||
| AK297130.1 | ||||
| BC007256.1 | 1 | 184 | AAH07256.1 | |
| BC048974.1 | AAH48974.1 | |||
| DA530642.1 | 1 | 193 | ||
| DB160047.1 |
More Information
Set Membership: |
|

Genomic Map