Assay Details
Target Gene Details
Entrez Gene ID: | 29992 |
Gene Name: | paired immunoglobin like type 2 receptor alpha |
Gene Aliases: |
FDF03 |
Location: |
Chr.7:100373445-100400099 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PILRA | NM_013439.2 | 3 | 745 | NP_038467.2 |
| NM_178272.1 | NP_840056.1 | |||
| NM_178273.1 | NP_840057.1 | |||
| AF161080.1 | 3 | 745 | AAD52964.1 | |
| AJ400841.1 | 3 | 686 | CAC01613.1 | |
| AJ400842.1 | CAC01614.1 | |||
| AJ400843.1 | CAC01615.1 | |||
| BC017812.1 | AAH17812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
| nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map