Assay Details
Target Gene Details
Entrez Gene ID: | 5715 |
Gene Name: | proteasome 26S subunit, non-ATPase 9 |
Gene Aliases: |
Rpn4, p27 |
Location: |
Chr.12:121888731-121917865 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMD9 | NM_001261400.2 | NP_001248329.1 | ||
| NM_002813.6 | 3 | 469 | NP_002804.2 | |
| NR_048555.2 | ||||
| AK315389.1 | 3 | 438 | ||
| BC002383.2 | 3 | 429 | AAH02383.1 | |
| BC004184.2 | 3 | 429 | AAH04184.1 | |
| BC004213.2 | 3 | 429 | AAH04213.1 | |
| BI832046.1 | 3 | 463 | ||
| BM850893.1 | ||||
| BU956605.1 | 3 | 450 | ||
| CB111716.1 | ||||
| CN260241.1 | 3 | 475 |
More Information
Set Membership: |
|

Genomic Map