Assay Details
Target Gene Details
Entrez Gene ID: | 118 |
Gene Name: | adducin 1 |
Gene Aliases: |
ADDA |
Location: |
Chr.4:2843732-2930076 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADD1 | NM_001119.4 | 2 | 330 | NP_001110.2 |
| NM_001286645.1 | 3 | 1005 | NP_001273574.1 | |
| NM_014189.3 | 2 | 330 | NP_054908.2 | |
| NM_014190.3 | 2 | 330 | NP_054909.2 | |
| NM_176801.2 | 2 | 330 | NP_789771.1 | |
| XM_005247933.2 | 2 | 455 | XP_005247990.1 | |
| XM_005247934.2 | 2 | 419 | XP_005247991.1 | |
| XM_005247935.2 | 2 | 455 | XP_005247992.1 | |
| XM_005247936.2 | 2 | 455 | XP_005247993.1 | |
| XM_005247937.2 | 2 | 455 | XP_005247994.1 | |
| XM_017007703.1 | 2 | 180 | XP_016863192.1 | |
| XM_017007704.1 | 2 | 419 | XP_016863193.1 | |
| XM_017007705.1 | 2 | 419 | XP_016863194.1 | |
| XM_017007706.1 | 2 | 419 | XP_016863195.1 | |
| XM_017007707.1 | 1 | 177 | XP_016863196.1 | |
| XM_017007708.1 | 2 | 455 | XP_016863197.1 | |
| XM_017007709.1 | 2 | 419 | XP_016863198.1 | |
| XM_017007710.1 | 1 | 177 | XP_016863199.1 | |
| AK295457.1 | 2 | 417 | ||
| AK309186.1 | 2 | 208 | ||
| BC013393.2 | 2 | 265 | AAH13393.1 | |
| BC042998.1 | 3 | 1010 | AAH42998.1 | |
| BI460192.1 | 2 | 353 | ||
| CR995783.1 | 2 | 305 | ||
| D44632.1 | 2 | 347 | ||
| L07261.1 | 1 | 144 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv313n21 | Chr.4:2845072 - 2879529 on Build GRCh38 | Gain |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv593435 | Chr.4:2769045 - 2879529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map