Assay Details
Target Gene Details
Entrez Gene ID: | 3400 |
Gene Name: | inhibitor of DNA binding 4, HLH protein |
Gene Aliases: |
IDB4, bHLHb27 |
Location: |
Chr.6:19837370-19842200 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ID4 | NM_001546.3 | NP_001537.1 | ||
| BC014941.1 | AAH14941.1 | |||
| U16153.1 | AAA82882.1 | |||
| U28368.1 | AAA73923.1 | |||
| Y07958.1 | CAA69255.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv601089 | Chr.6:19837856 - 19839050 on Build GRCh38 | Loss |
|
| esv23665 | Chr.6:19837558 - 19839541 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map