Assay Details
Target Gene Details
Entrez Gene ID: | 9513 |
Gene Name: | FMR1 autosomal homolog 2 |
Gene Aliases: |
FMR1L2, FXR2P |
Location: |
Chr.17:7591230-7614897 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FXR2 | NM_004860.3 | NP_004851.2 | ||
| AK226082.1 | ||||
| AK313836.1 | ||||
| BC020090.1 | AAH20090.1 | |||
| BC051907.1 | AAH51907.1 | |||
| BC067272.1 | AAH67272.1 | |||
| BT009817.1 | AAP88819.1 | |||
| U31501.1 | AAC50292.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map