Assay Details
Target Gene Details
Entrez Gene ID: | 6722 |
Gene Name: | serum response factor |
Gene Aliases: |
MCM1 |
Location: |
Chr.6:43171295-43181506 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRF | NM_001292001.1 | NP_001278930.1 | ||
| NM_003131.3 | NP_003122.1 | |||
| AB209128.1 | 2 | 3440 | BAD92365.1 | |
| AK300467.1 | ||||
| BC048211.1 | AAH48211.1 | |||
| BC052572.1 | AAH52572.1 | |||
| J03161.1 | AAA36647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3571027 | Chr.6:43159598 - 43178415 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
| nsv521730 | Chr.6:43121691 - 43216394 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map