Assay Details
Target Gene Details
Entrez Gene ID: | 79693 |
Gene Name: | yrdC N6-threonylcarbamoyltransferase domain containing |
Gene Aliases: |
DRIP3, IRIP, SUA5 |
Location: |
Chr.1:37802942-37808193 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YRDC | NM_024640.3 | NP_078916.3 | ||
| AF250397.1 | AAQ14263.1 | |||
| AK027129.1 | BAB15668.1 | |||
| AL524812.3 | ||||
| AY172561.1 | AAO41711.1 | |||
| AY286019.1 | AAP37053.1 | |||
| AY286020.1 | AAP37054.1 | |||
| BC008984.1 | ||||
| BC067857.1 | AAH67857.2 | |||
| BC068057.1 | ||||
| BC080186.1 | AAH80186.2 |
More Information
Set Membership: |
|

Genomic Map