Assay Details
Target Gene Details
Entrez Gene ID: | 11083 |
Gene Name: | death inducer-obliterator 1 |
Gene Aliases: |
BYE1, C20orf158, DATF-1, DATF1, DIDO2, DIDO3, DIO-1, DIO1, dJ885L7.8 |
Location: |
Chr.20:62877738-62937952 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIDO1 | NM_001193369.1 | 16 | 7387 | NP_001180298.1 |
| NM_033081.2 | 16 | 7431 | NP_149072.2 | |
| XM_011528504.1 | 17 | 7472 | XP_011526806.1 | |
| XM_011528505.1 | 17 | 7539 | XP_011526807.1 | |
| XM_011528506.1 | 17 | 14323 | XP_011526808.1 | |
| XM_011528507.1 | 17 | 7407 | XP_011526809.1 | |
| XM_011528508.1 | 17 | 7548 | XP_011526810.1 | |
| AL133063.1 | 1 | 2211 | CAB61387.1 | |
| AY481572.1 | 16 | 7339 | AAS49899.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828794 | Chr.20:62832958 - 62880035 on Build GRCh38 | Loss |
|
| nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
|
| nsv428380 | Chr.20:62632018 - 62935383 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map