Assay Details
Target Gene Details
Entrez Gene ID: | 55974 |
Gene Name: | solute carrier family 50 member 1 |
Gene Aliases: |
HsSWEET1, RAG1AP1, SCP, SWEET1, slv |
Location: |
Chr.1:155135253-155138858 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC50A1 | NM_001122837.1 | NP_001116309.1 | ||
| NM_001122839.1 | 3 | 340 | NP_001116311.1 | |
| NM_001287586.1 | 2 | 243 | NP_001274515.1 | |
| NM_001287587.1 | 3 | 360 | NP_001274516.1 | |
| NM_001287588.1 | NP_001274517.1 | |||
| NM_001287589.1 | NP_001274518.1 | |||
| NM_001287590.1 | NP_001274519.1 | |||
| NM_001287591.1 | 3 | 415 | NP_001274520.1 | |
| NM_001287592.1 | NP_001274521.1 | |||
| NM_018845.3 | 3 | 340 | NP_061333.2 | |
| XM_005245339.4 | 3 | 526 | XP_005245396.1 | |
| XM_005245340.3 | 3 | 607 | XP_005245397.1 | |
| XM_005245341.4 | 3 | 558 | XP_005245398.1 | |
| XM_005245342.3 | 2 | 166 | XP_005245399.1 | |
| XM_006711453.2 | 3 | 340 | XP_006711516.1 | |
| AF126023.1 | 3 | 300 | AAF17232.1 | |
| AF126024.1 | AAF17233.1 | |||
| AK289551.1 | 3 | 341 | ||
| BC005943.1 | 3 | 284 | AAH05943.1 | |
| BC009621.1 | 3 | 302 | AAH09621.1 | |
| BI195885.1 | ||||
| BM806886.1 | 3 | 402 | ||
| BP248307.1 | 3 | 485 | ||
| BU160680.1 | 3 | 282 | ||
| BU564973.1 | ||||
| BX093804.1 | ||||
| CR457148.1 | 3 | 240 | CAG33429.1 | |
| DA113832.1 | 3 | 341 | ||
| X84712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| nsv547961 | Chr.1:155109822 - 155225189 on Build GRCh38 | Loss |
|
| nsv998756 | Chr.1:155107614 - 155164069 on Build GRCh38 | Gain |
|
| esv3587615 | Chr.1:155047853 - 155175013 on Build GRCh38 | Gain |
|
| dgv3n68 | Chr.1:155122502 - 155343618 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map